Tetrabrombisphenol A, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4,4'-(Propane-2,2-diyl)bis(2,6-dibromophenol), 3,3',5,5'-Tetrabromobisphenol A

M-Clarity™ Qualitätsstufe = MQ100

330396-100G
SIAL330396-100GEA 80.1 EUR
SIAL330396-100G
Tetrabrombisphenol A, Sigma-Aldrich®
Tetrabrombisphenol A
Formel: C₁₅H₁₂Br₄O₂
Molecular Weight: 543.87 g/mol
MDL: MFCD00013962
CAS-Nummer: 79-94-7
EINECS: 201-236-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR