N-Phenylglycin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Anilinoacetic acid, 2-(Phenylamino)acetic acid

M-Clarity™ Qualitätsstufe = MQ200

330469-25G
SIAL330469-25GEA 62.4 EUR
SIAL330469-25G
N-Phenylglycin, Sigma-Aldrich®
N-Phenylglycin
Formel: C₈H₉NO₂
Molecular Weight: 151,17 g/mol
MDL: MFCD00014009
CAS-Nummer: 103-01-5
EINECS: 203-070-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR