(R)-(-)-2-Pentanol, Sigma-Aldrich®

Lieferant: Merck
Gefahr

Synonyme: (R)-(-)-Propyl methyl carbinol, (R)-(-)-2-Hydroxy pentane, (R)-(-)-Methyl propyl carbinol

M-Clarity™ Qualitätsstufe = MQ200

330523-500MG
SIAL330523-500MGEA 162 EUR
SIAL330523-500MG
(R)-(-)-2-Pentanol, Sigma-Aldrich®
(R)-(-)-2-Pentanol
Formel: C₅H₁₂O
Molecular Weight: 88.15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00065953
CAS-Nummer: 31087-44-2
UN: 1105
ADR: 3 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR