α,α,α,6-Tetrafluor-o-toluylsäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

330809-5G
SIAL330809-5GEA 78 EUR
SIAL330809-5G
α,α,α,6-Tetrafluor-o-toluylsäure, Sigma-Aldrich®
α,α,α,6-Tetrafluor-o-toluylsäure
Formel: C₈H₄F₄O₂
Molecular Weight: 208,11 g/mol
MDL: MFCD00040981
CAS-Nummer: 32890-94-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR