4-Cyan-4'-heptylbiphenyl, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Cyano-4'-n-heptylbiphenyl, 4'-Heptyl-[1,1'-biphenyl]-4-carbonitrile

M-Clarity™ Qualitätsstufe = MQ100

330817-1G
SIAL330817-1GEA 104 EUR
SIAL330817-1G
4-Cyan-4'-heptylbiphenyl, Sigma-Aldrich®
4-Cyan-4'-heptylbiphenyl
Formel: C₂₀H₂₃N
Molecular Weight: 277,41 g/mol
MDL: MFCD00074915
CAS-Nummer: 41122-71-8
EINECS: 255-229-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR