Pentafluoriodethan
Lieferant: Merck
Synonyme:
Perfluorethyliodid, Iodpentafluorethan
M-Clarity™ Qualitätsstufe = MQ100
331015-300G
SIAL331015-300GEA
0
EUR
SIAL331015-300G
Pentafluoriodethan
Pentafluoriodethan
Formel:
C₂F₅I Molecular Weight: 245.92 g/mol Schmelzpunkt: –92 °C Dichte: 2.1605 g/cm³ (0 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00039400 CAS-Nummer: 354-64-3 EINECS: 206-566-7 UN: 1956 ADR: 2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...