3,5-Dimethyl-1H-pyrazol-1-methanol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3,5-Dimethyl-1-(hydroxymethyl)-1H-pyrazole

M-Clarity™ Qualitätsstufe = MQ100

331457-25G
SIAL331457-25GEA 76.8 EUR
SIAL331457-25G
3,5-Dimethyl-1H-pyrazol-1-methanol, Sigma-Aldrich®
3,5-Dimethyl-1H-pyrazol-1-methanol
Formel: C₆H₁₀N₂O
MDL: MFCD00020738
CAS-Nummer: 85264-33-1
EINECS: 286-553-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR