Tetramethylammoniumhydroxid 25% aqueous solution, Sigma-Aldrich®

Lieferant: Merck
Gefahr


MQ200

331635-1L 331635-250ML
SIAL331635-1LEA 194 EUR
SIAL331635-1L SIAL331635-250ML
Tetramethylammoniumhydroxid 25% aqueous solution, Sigma-Aldrich®
Tetramethylammonium hydroxide
Formel: C₄H₁₃NO
Molecular Weight: 91.15 g/mol
Dichte: 1.02 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00008280
CAS-Nummer: 75-59-2
EINECS: 200-882-9
UN: 1835
ADR: 8 II
Merck Index: 14,09224

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR