DL-tert-Leucin (H-DL-Tle-OH), Sigma-Aldrich®

Lieferant: Merck

Synonyme: H-DL-Tle-OH

M-Clarity™ Qualitätsstufe = MQ200

332178-500MG
SIAL332178-500MGEA 96 EUR
SIAL332178-500MG
DL-tert-Leucin (H-DL-Tle-OH), Sigma-Aldrich®
DL-tert-Leucin (H-DL-Tle-OH)
Formel: C₆H₁₃NO₂
Molecular Weight: 131,2 g/mol
MDL: MFCD00065933
CAS-Nummer: 33105-81-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR