3-Thiophenmethanol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Thiophen-3-methanol, 3-Thienylmethanol, 3-(Hydroxymethyl)thiophen

M-Clarity™ Qualitätsstufe = MQ100

332399-5G
SIAL332399-5GEA 82.3 EUR
SIAL332399-5G
3-Thiophenmethanol, Sigma-Aldrich®
3-Thiophenmethanol
Formel: C₅H₆OS
Molecular Weight: 114,17 g/mol
MDL: MFCD00014534
CAS-Nummer: 71637-34-8
EINECS: 275-741-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR