Graphit, Sigma-Aldrich®
Lieferant: Merck
332461-12KG
332461-2.5KG
SIAL332461-12KGEA
270
EUR
SIAL332461-12KG
SIAL332461-2.5KG
Graphit, Sigma-Aldrich®
Graphit
Formel:
C Schmelzpunkt: 3827 °C Dichte: 2,09…2.23 g/cm³ (20 °C) |
MDL:
MFCD00144065 CAS-Nummer: 7782-42-5 EINECS: 231-955-3 Merck Index: 12,04560 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...