Dibenzo-30-krone-10 (Dibenzo-1,4,7,10,13,16,19,22,25,28-decaoxacyclotriacontan), Sigma-Aldrich®

Lieferant: Merck

Synonyme: 6,7,9,10,12,13,15,16,23,24,26,27,29,30,32,33-Hexadecahydrodibenzo[b,q][1,4,7,10,13,16,19,22,25,28]decaoxacyclotriacontine, Dibenzo-30-crown-10-ether

M-Clarity™ Qualitätsstufe = MQ100

332518-250MG
SIAL332518-250MGEA 92.6 EUR
SIAL332518-250MG
Dibenzo-30-krone-10 (Dibenzo-1,4,7,10,13,16,19,22,25,28-decaoxacyclotriacontan), Sigma-Aldrich®
Dibenzo-30-krone-10 (Dibenzo-1,4,7,10,13,16,19,22,25,28-decaoxacyclotriacontan)
Formel: C₂₈H₄₀O₁₀
Molecular Weight: 536.62 g/mol
MDL: MFCD00054624
CAS-Nummer: 17455-25-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR