2-Methyl-3-indolylessigsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Methylindole-3-acetic acid

M-Clarity™ Qualitätsstufe = MQ100

333301-5G
SIAL333301-5GEA 80.1 EUR
SIAL333301-5G
2-Methyl-3-indolylessigsäure, Sigma-Aldrich®
2-Methyl-3-indolylessigsäure
Formel: C₁₁H₁₁NO₂
Molecular Weight: 189,21 g/mol
MDL: MFCD00075006
CAS-Nummer: 1912-43-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR