Chrom(III)fluorid Tetrahydrat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Chromtrifluorid Tetrahydrat

M-Clarity™ Qualitätsstufe = MQ100

333387-100G
SIAL333387-100GEA 113 EUR
SIAL333387-100G
Chrom(III)fluorid Tetrahydrat, Sigma-Aldrich®
Chrom(III)fluorid Tetrahydrat
Formel: CrF₃·4H₂O
MDL: MFCD00016045
CAS-Nummer: 123333-98-2
EINECS: 232-137-9
UN: 1756
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR