Methyl-4-(trifluormethyl)benzoat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Methyl 4-(trifluoromethyl)benzoate
M-Clarity™ Qualitätsstufe = MQ100
333441-5G
SIAL333441-5GEA
53
EUR
SIAL333441-5G
Methyl-4-(trifluormethyl)benzoat, Sigma-Aldrich®
Methyl-4-(trifluormethyl)benzoat
Formel:
C₉H₇F₃O₂ Molecular Weight: 204,15 g/mol |
MDL:
MFCD00042324 CAS-Nummer: 2967-66-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...