Titan(IV)sulfid, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
Titandisulfid
M-Clarity™ Qualitätsstufe = MQ100
333492-10G
SIAL333492-10GEA
338
EUR
SIAL333492-10G
Titan(IV)sulfid, Sigma-Aldrich®
Titan(IV)sulfid
Formel:
S₂Ti Molecular Weight: 112,01 g/mol Dichte: 3,22 g/cm³ (20 °C) |
MDL:
MFCD00049598 CAS-Nummer: 12039-13-3 EINECS: 234-883-0 UN: 3134 ADR: 4.3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...