1,4-Naphthalindicarbonsäure, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
M-Clarity™ Qualitätsstufe = MQ100
333581-1G
SIAL333581-1GEA
0
EUR
SIAL333581-1G
1,4-Naphthalindicarbonsäure, Sigma-Aldrich®
1,4-Naphthalindicarbonsäure
Formel:
C₁₂H₈O₄ |
MDL:
MFCD00014312 CAS-Nummer: 605-70-9 EINECS: 210-094-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...