Iridium(IV)chlorid Hydrat, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
334480-500MG
SIAL334480-500MGEA
144
EUR
SIAL334480-500MG
Iridium(IV)chlorid Hydrat, Sigma-Aldrich®
Iridium(IV)chlorid Hydrat
Formel:
Cl₄Ir·xH₂O |
MDL:
MFCD00150243 CAS-Nummer: 207399-11-9 EINECS: 233-048-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...