1,2-Diiodtetrafluorbenzol, Sigma-Aldrich®

Lieferant: Merck
Achtung


M-Clarity™ Qualitätsstufe = MQ200

334707-5G
SIAL334707-5GEA 148 EUR
SIAL334707-5G
1,2-Diiodtetrafluorbenzol, Sigma-Aldrich®
1,2-Diiodtetrafluorbenzol
Formel: C₆F₄I₂
Molecular Weight: 401,87 g/mol
MDL: MFCD00019011
CAS-Nummer: 2708-97-6
EINECS: 220-303-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR