2-Fluor-4-nitroanisol, Sigma-Aldrich®

Lieferant: Merck
Achtung


M-Clarity™ Qualitätsstufe = MQ100

334863-5G
SIAL334863-5GEA 32.1 EUR
SIAL334863-5G
2-Fluor-4-nitroanisol, Sigma-Aldrich®
2-Fluor-4-nitroanisol
Formel: C₇H₆FNO₃
Molecular Weight: 171,13 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00061095
CAS-Nummer: 455-93-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR