3-Fluor-p-anisidin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Amino-2-fluoranisol, 3-Fluor-4-methoxyanilin, 3-Fluor-4-anisidin

M-Clarity™ Qualitätsstufe = MQ100

334871-5G
SIAL334871-5GEA 85.3 EUR
SIAL334871-5G
3-Fluor-p-anisidin, Sigma-Aldrich®
3-Fluor-p-anisidin
Formel: C₇H₈FNO
Molecular Weight: 141,15 g/mol
MDL: MFCD00075040
CAS-Nummer: 366-99-4
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR