2,5-Dimethyl-p-phenylendiamin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

336068-5G
SIAL336068-5GEA 59.4 EUR
SIAL336068-5G
2,5-Dimethyl-p-phenylendiamin, Sigma-Aldrich®
2,5-Dimethyl-p-phenylendiamin
Formel: C₈H₁₂N₂
MDL: MFCD00052675
CAS-Nummer: 6393-01-7
EINECS: 229-003-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR