Iridium(III)chlorid, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
336807-2G
336807-500MG
SIAL336807-2GEA
616
EUR
SIAL336807-2G
SIAL336807-500MG
Iridium(III)chlorid, Sigma-Aldrich®
Iridium(III)chlorid
Formel:
Cl₃Ir Dichte: 5,3 g/cm³ (20 °C) |
MDL:
MFCD00011063 CAS-Nummer: 10025-83-9 EINECS: 233-044-6 UN: 3260 ADR: 8,II Merck Index: 12,05101 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...