1,3,5-Tribenzylhexahydro-1,3,5-triazin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1,3,5-Tribenzyl-1,3,5-triazinane

337722-25G
SIAL337722-25GEA 137 EUR
SIAL337722-25G
1,3,5-Tribenzylhexahydro-1,3,5-triazin, Sigma-Aldrich®
1,3,5-Tribenzylhexahydro-1,3,5-triazin
Formel: C₂₄H₂₇N₃
Molecular Weight: 357,5 g/mol
MDL: MFCD00014599
CAS-Nummer: 2547-66-2
EINECS: 219-831-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR