4-Chlor-2,6-dinitroanilin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

337854-5G
SIAL337854-5GEA 0 EUR
SIAL337854-5G
4-Chlor-2,6-dinitroanilin, Sigma-Aldrich®
4-Chlor-2,6-dinitroanilin
Formel: C₆H₄ClN₃O₄
Molecular Weight: 217,57 g/mol
CAS-Nummer: 5388-62-5
EINECS: 226-381-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR