(R)-(-)-Glycidylbutyrat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (-)-[(2R)-Oxiran-2-yl]methyl butanoate, (R)-Oxiran-2-ylmethyl butyrate

M-Clarity™ Qualitätsstufe = MQ100

338125-5G
SIAL338125-5GEA 37 EUR
SIAL338125-5G
(R)-(-)-Glycidylbutyrat, Sigma-Aldrich®
(R)-(-)-Glycidylbutyrat
Formel: C₇H₁₂O₃
Molecular Weight: 144,17 g/mol
MDL: MFCD00075120
CAS-Nummer: 60456-26-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR