Triphenylphosphoniumbromid, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ200
338389-5G
SIAL338389-5GEA
35.4
EUR
SIAL338389-5G
Triphenylphosphoniumbromid, Sigma-Aldrich®
Triphenylphosphonium bromide
Formel:
C₁₈H₁₆BrP |
MDL:
MFCD00035107 CAS-Nummer: 6399-81-1 EINECS: 229-012-6 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...