Isochinolin-3-carbonitril, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Isoquinolinecarbonitrile

M-Clarity™ Qualitätsstufe = MQ100

338532-1G
SIAL338532-1GEA 186 EUR
SIAL338532-1G
Isochinolin-3-carbonitril, Sigma-Aldrich®
Isochinolin-3-carbonitril
Formel: C₁₀H₆N₂
Molecular Weight: 154.17 g/mol
MDL: MFCD00075136
CAS-Nummer: 26947-41-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR