4-Brom-3-nitroanisol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-Bromo-4-methoxy-2-nitrobenzene

M-Clarity™ Qualitätsstufe = MQ200

338737-5G
SIAL338737-5GEA 28.4 EUR
SIAL338737-5G
4-Brom-3-nitroanisol, Sigma-Aldrich®
4-Brom-3-nitroanisol
Formel: C₇H₆BrNO₃
Molecular Weight: 232.03 g/mol
MDL: MFCD00051511
CAS-Nummer: 5344-78-5
EINECS: 226-290-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR