Eisen(II)acetat, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
339199-50G
SIAL339199-50GEA
580
EUR
SIAL339199-50G
Eisen(II)acetat, Sigma-Aldrich®
Eisen(II)acetat
Formel:
C₄H₆FeO₄ Molecular Weight: 173,94 g/mol Schmelzpunkt: 190 °C |
MDL:
MFCD00058909 CAS-Nummer: 3094-87-9 EINECS: 221-441-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...