Mangan(III)fluorid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

339296-10G
SIAL339296-10GEA 108 EUR
SIAL339296-10G
Mangan(III)fluorid, Sigma-Aldrich®
Mangan(III)fluorid
Formel: F₃Mn
Dichte: 3,54 g/cm³ (20 °C)
MDL: MFCD00016223
CAS-Nummer: 7783-53-1
EINECS: 232-006-6
UN: 3288
ADR: 6.1,III
Merck Index: 12,05784

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR