2-Amino-4-(4-chlorphenyl)thiazol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-(4-Chlorophenyl)thiazol-2-amine

M-Clarity™ Qualitätsstufe = MQ100

339369-1G
SIAL339369-1GEA 34.6 EUR
SIAL339369-1G
2-Amino-4-(4-chlorphenyl)thiazol, Sigma-Aldrich®
2-Amino-4-(4-chlorophenyl)thiazole
Formel: C₉H₇ClN₂S
Molecular Weight: 210,69 g/mol
MDL: MFCD00047058
CAS-Nummer: 2103-99-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR