Nickel(II)carbonat basisch Tetrahydrat, Sigma-Aldrich®

Lieferant: Merck

339776-500G
SIAL339776-500GEA 145 EUR
SIAL339776-500G
Nickel(II)carbonat basisch Tetrahydrat, Sigma-Aldrich®
Nickel(II)carbonat basisch Tetrahydrat
Formel: C₂H₆Ni₅O₁₂·4H₂O
CAS-Nummer: 12244-51-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR