1,5-Dihydroxy-9,10-anthrachinon, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: 1,5-Dihydroxyanthrachinon

340014-100G
SIAL340014-100GEA 144 EUR
SIAL340014-100G
1,5-Dihydroxy-9,10-anthrachinon, Sigma-Aldrich®
1,5-Dihydroxy-9,10-anthrachinon
Formel: C₁₄H₈O₄
Molecular Weight: 240,22 g/mol
MDL: MFCD00001210
CAS-Nummer: 117-12-4
EINECS: 204-175-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR