(4S,5R)-(-)-4-Methyl-5-phenyl-2-oxazolidinon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (4S,5R)-4-Methyl-5-phenyloxazolidin-2-one, (4S,5R)-(-)-4-Methyl-5-phenyloxazolidin-2-one

M-Clarity™ Qualitätsstufe = MQ100

340529-5G
SIAL340529-5GEA 70.6 EUR
SIAL340529-5G
(4S,5R)-(-)-4-Methyl-5-phenyl-2-oxazolidinon, Sigma-Aldrich®
(4S,5R)-(-)-4-Methyl-5-phenyl-2-oxazolidinon
Formel: C₁₀H₁₁NO₂
Molecular Weight: 177,2000 g/mol
MDL: MFCD00066226
CAS-Nummer: 16251-45-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR