3-Brom-4-fluorbenzoesäure, Sigma-Aldrich®

Lieferant: Merck
Achtung


M-Clarity™ Qualitätsstufe = MQ100

341355-1G
SIAL341355-1GEA 26 EUR
SIAL341355-1G
3-Brom-4-fluorbenzoesäure, Sigma-Aldrich®
3-Bromo-4-fluorobenzoic acid
Formel: C₇H₄BrFO₂
Molecular Weight: 219,01 g/mol
MDL: MFCD00042463
CAS-Nummer: 1007-16-5
EINECS: 213-751-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR