2,3-Difluor-6-nitrophenol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1,2-Difluoro-3-hydroxy-4-nitrobenzene

M-Clarity™ Qualitätsstufe = MQ200

341533-5G
SIAL341533-5GEA 81.2 EUR
SIAL341533-5G
2,3-Difluor-6-nitrophenol, Sigma-Aldrich®
2,3-Difluor-6-nitrophenol
Formel: C₆H₃F₂NO₃
Molecular Weight: 175,09 g/mol
Schmelzpunkt: 61 °C
MDL: MFCD00042255
CAS-Nummer: 82419-26-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR