Calcium iodate 98%, Sigma-Aldrich®

Lieferant: Merck
Gefahr


M-Clarity™ Qualitätsstufe = MQ100

341606-100G
SIAL341606-100GEA 64.5 EUR
SIAL341606-100G
Calcium iodate 98%, Sigma-Aldrich®
Calcium iodate
Formel: CaI₂O₆
Molecular Weight: 389,881 g/mol
Dichte: 4,520 g/cm³
MDL: MFCD00036265
CAS-Nummer: 7789-80-2
EINECS: 232-191-3
UN: 1479
ADR: 5.1,II

Bestellen

Spezifikation Testergebnisse

Appearance (Colour) White to Yellow
Appearance (Form) Powder
X-Ray Diffraction Conforms to Structure
Titration by Na₂S₂O₃ (% IO3) 87.5 - 92.0%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR