Bromcyan 3 mol/l in Dichlormethan, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Cyanogenbromid

M-Clarity™ Qualitätsstufe = MQ100

341894-100ML
SIAL341894-100MLEA 82.6 EUR
SIAL341894-100ML
Bromcyan 3 mol/l in Dichlormethan, Sigma-Aldrich®
Bromcyan
Formel: CBrN
Molecular Weight: 105,92 g/mol
MDL: MFCD00011597
CAS-Nummer: 506-68-3
EINECS: 208-051-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR