3-(1,1,2,2-Tetrafluorethoxy)anilin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

341983-1G
SIAL341983-1GEA 113 EUR
SIAL341983-1G
3-(1,1,2,2-Tetrafluorethoxy)anilin, Sigma-Aldrich®
3-(1,1,2,2-Tetrafluorethoxy)anilin
Formel: C₈H₇F₄NO
MDL: MFCD00042457
CAS-Nummer: 831-75-4
EINECS: 212-610-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR