4-(1,1,2,2-Tetrafluorethoxy)benzaldehyd, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

342009-1G
SIAL342009-1GEA 57.6 EUR
SIAL342009-1G
4-(1,1,2,2-Tetrafluorethoxy)benzaldehyd, Sigma-Aldrich®
4-(1,1,2,2-Tetrafluoroethoxy)benzaldehyde
Formel: C₉H₆F₄O₂
Molecular Weight: 222,1364 g/mol
MDL: MFCD00039577
CAS-Nummer: 35295-36-4
EINECS: 252-497-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR