5-(Trifluormethoxy)salicylaldehyd, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Hydroxy-5-(trifluoromethoxy)benzaldehyde

M-Clarity™ Qualitätsstufe = MQ100

342157-500MG
SIAL342157-500MGEA 0 EUR
SIAL342157-500MG
5-(Trifluormethoxy)salicylaldehyd, Sigma-Aldrich®
5-(Trifluormethoxy)salicylaldehyd
Formel: C₈H₅F₃O₃
Molecular Weight: 206,12 g/mol
MDL: MFCD00075249
CAS-Nummer: 93249-62-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR