2-Amino-6-chlorpurin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
6-Chlor-7H-purin-2-ylamin
M-Clarity™ Qualitätsstufe = MQ100
342300-1G
342300-5G
SIAL342300-1GEA
37.8
EUR
SIAL342300-1G
SIAL342300-5G
2-Amino-6-chlorpurin, Sigma-Aldrich®
2-Amino-6-chlorpurin
Formel:
C₅H₄ClN₅ Molecular Weight: 169.57 g/mol Schmelzpunkt: 300 |
MDL:
MFCD00075252 CAS-Nummer: 10310-21-1 EINECS: 233-686-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...