3-Chlor-6-phenylpyridazin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Phenyl-6-chloropyridazine

M-Clarity™ Qualitätsstufe = MQ100

342319-1G
SIAL342319-1GEA 47.9 EUR
SIAL342319-1G
3-Chlor-6-phenylpyridazin, Sigma-Aldrich®
3-Chloro-6-phenylpyridazine
Formel: C₁₀H₇ClN₂
Molecular Weight: 190,6300 g/mol
MDL: MFCD00075253
CAS-Nummer: 20375-65-9
EINECS: 243-772-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR