Cobalt(II)hydroxid, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
M-Clarity™ Qualitätsstufe = MQ200
342440-250G
SIAL342440-250GEA
72.9
EUR
SIAL342440-250G
Cobalt(II)hydroxid, Sigma-Aldrich®
Cobalt(II)hydroxid
Formel:
H₂CoO₂ Dichte: 3,597 g/cm³ (15 °C) |
MDL:
MFCD00016015 CAS-Nummer: 21041-93-0 EINECS: 244-166-4 Merck Index: 13,02467 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...