Nickelsulfid, Sigma-Aldrich®
Lieferant: Merck
|
Gefahr
|
Synonyme:
Nickel subsulphide, Nickel disulphane
M-Clarity™ Qualitätsstufe = MQ100
343226-25G
SIAL343226-25GEA
297
EUR
SIAL343226-25G
Nickelsulfid, Sigma-Aldrich®
Nickelsulfid
Formel:
Ni₃S₂ Molecular Weight: 240,21 g/mol |
MDL:
MFCD00217044 CAS-Nummer: 12035-72-2 EINECS: 234-829-6 UN: 3077 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...