2-Chlor-p-benzochinon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Chlorocyclohexa-2,5-diene-1,4-dione, 2-Chloro-1,4-benzoquinone

M-Clarity™ Qualitätsstufe = MQ100

343382-5G
SIAL343382-5GEA 98.1 EUR
SIAL343382-5G
2-Chlor-p-benzochinon, Sigma-Aldrich®
2-Chlor-p-benzochinon
Formel: C₆H₃ClO₂
Molecular Weight: 142,54 g/mol
MDL: MFCD00075260
CAS-Nummer: 695-99-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR