5-Isopropyl-1,3-cyclohexandion ≥99% (durch GC), Sigma-Aldrich®
Lieferant: Merck
Synonyme:
5-Isopropylcyclohexane-1,3-dione
M-Clarity™ Qualitätsstufe = MQ100
343447-5G
SIAL343447-5GEA
143
EUR
SIAL343447-5G
5-Isopropyl-1,3-cyclohexandion ≥99% (durch GC), Sigma-Aldrich®
5-Isopropyl-1,3-cyclohexandion
5-Isopropyl-1,3-cyclohexanedione may be used as starting reagent for the synthesis of carvotanacetone analogs. It may be used in the preparation of monocyclic terepenes. It may be used in the synthesis of 2H-pyrans, via reaction with α,β-unsaturated aldehydes.
Formel:
C₉H₁₄O₂·H₂O Molecular Weight: 154,21 g/mol (anhy) |
MDL:
MFCD00075263 CAS-Nummer: 18456-87-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...