1-Boc-piperazin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: tert-Butyl 1-piperazinecarboxylate, tert-butyl piperazine-1-carboxylate

M-Clarity™ Qualitätsstufe = MQ200

343536-1G 343536-25G 343536-5G
SIAL343536-1GEA 32.4 EUR
SIAL343536-1G SIAL343536-25G SIAL343536-5G
1-Boc-piperazin, Sigma-Aldrich®
1-Boc-piperazin
Formel: C₉H₁₈N₂O₂
MDL: MFCD00075265
CAS-Nummer: 57260-71-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR