1-(2-Naphthoyl)-3,3,3-trifluoraceton, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4,4,4-Trifluoro-1-(naphthalen-2-yl)butane-1,3-dione, 4,4,4-Trifluoro-1-(2-naphthyl)-1,3-butanedione

M-Clarity™ Qualitätsstufe = MQ200

343633-5G
SIAL343633-5GEA 38.9 EUR
SIAL343633-5G
1-(2-Naphthoyl)-3,3,3-trifluoraceton, Sigma-Aldrich®
1-(2-Naphthoyl)-3,3,3-trifluoraceton
Formel: C₁₄H₉F₃O₂
Molecular Weight: 266.22 g/mol
MDL: MFCD00054501
CAS-Nummer: 893-33-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR